Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX26B Antibody, Novus Biologicals™
SDP

Catalog No. NBP18020520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18020520 20 μL
NBP180205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18020520 Supplier Novus Biologicals Supplier No. NBP18020520UL

Rabbit Polyclonal Antibody

DDX26B Polyclonal specifically detects DDX26B in Human samples. It is validated for Western Blot.

Specifications

Antigen DDX26B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_872346
Gene Alias DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B
Gene Symbols DDX26B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human DDX26B. Peptide sequence ASTEPEQLGSVPTDESAITQMCEVTGGRSYCVRTQRMLNQCLESLVQKVQ.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203522
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.