Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX3 Antibody (11F11D10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP214848G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214848G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214848G Supplier Novus Biologicals Supplier No. NBP214848G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDX3 Monoclonal specifically detects DDX3 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen DDX3
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 11F11D10
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry
Gene Accession No. O00571
Gene Alias DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X
Gene Symbols DDX3X
Host Species Mouse
Immunogen A partial recombinant protein containing amino acids 24-203 of human DDX3 was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1654
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.