Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX4/VASA Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DDX4/VASA |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5866
![]() ![]() |
Proteintech Group Inc
510421AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
DDX4 belongs to the DEAD box helicase family or DDX4/VASA subfamily. DEAD box proteins are characterized by the conserved Asp-Glu-Ala-Asp (DEAD) motif and are putative RNA helicases. This protein has a glycine-rich N terminus with multiple repeats of an RGG motif, which is believed to function in RNA binding. They have a role in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. The members of this family are involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a homolog of VASA proteins in Drosophila. It is specifically expressed in the germ cell lineage in both sexes and functions in germ cell development. It is required during gametogenesis in the adult and in the specification of the germ cell lineage during embryogenesis. This is conserved in invertebrate and vertebrate species.Specifications
DDX4/VASA | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q61496, Q9NQI0 | |
13206, 54514 | |
DDX4,VASA Fusion Protein Ag0447 | |
Primary | |
-20°C | |
DDX4 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DDX4, DEAD box protein 4, VASA, Vasa homolog | |
DDX4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title