Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX41 Antibody (PCRP-DDX41-1B4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB392118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392118 Supplier Novus Biologicals Supplier No. NBP308520R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDX41 Monoclonal antibody specifically detects DDX41 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen DDX41
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-DDX41-1B4
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias ABSDEAD-box protein abstrakt, DEAD (Asp-Glu-Ala-Asp) box polypeptide 41, DEAD box protein 41, DEAD box protein abstrakt homolog, EC 3.6.1, EC 3.6.4.13,2900024F02Rik, MGC8828, probable ATP-dependent RNA helicase DDX41, putative RNA helicase
Host Species Mouse
Immunogen Recombinant full-length human DDX41 protein (Uniprot: Q9UJV9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 51428
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.