Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX41 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501559115
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9115 Supplier Bethyl Laboratories, Inc Supplier No. A301050A
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. DDX41, also known as Abstrakt, interacts with and regulates the expression of sorting nexin-2 (SNX2), a protein involved in protein sorting in the trans-Golgi network. Recent evidence suggests that DDX41 also plays a role in the innate immune response by sensing intracellular viral DNA, triggering TBK1 and IRF3 activation, leading to a type I interferon immune response.

Specifications

Antigen DDX41
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DDX41
Gene Accession No. Q91VN6, Q9UJV9
Gene Alias ABS, 2900024F02Rik, DEAD box protein 41, DEAD box protein abstrakt homolog, DEAD-box protein abstrakt, probable ATP-dependent RNA helicase DDX41, putative RNA helicase
Gene Symbols Ddx41
Host Species Rabbit
Immunogen Between 572 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51428, 72935
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.