Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX46 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB127214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB127214 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB127214 Supplier Novus Biologicals Supplier No. NBP298581G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX46 Polyclonal specifically detects DDX46 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DDX46
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 46, DEAD box protein 46, EC 3.6.1, EC 3.6.4.13, FLJ25329, KIAA0801probable ATP-dependent RNA helicase DDX46, MGC9936, Prp5, PRP5 homolog, Prp5-like DEAD-box protein, PRPF5, RNA helicase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DDX46 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9879
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.