Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX49 Antibody, Novus Biologicals™
SDP

Catalog No. NBP169713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169713 Supplier Novus Biologicals Supplier No. NBP169713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX49 Polyclonal specifically detects DDX49 in Human samples. It is validated for Western Blot.

Specifications

Antigen DDX49
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9Y6V7
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 49, DEAD box protein 49, EC 3.6.1, EC 3.6.4.13, FLJ10432, probable ATP-dependent RNA helicase DDX49, R27090_2
Gene Symbols DDX49
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DDX49 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 49) The peptide sequence was selected from the N terminal of DDX49. Peptide sequence ELAYQIAEQFRVLGKPLGLKDCIIVGGMDMVAQALELSRKPHVVIATPGR.
Molecular Weight of Antigen 53 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54555
Test Specificity Yeast: 77%; Zebrafish: 77%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Canine, Equine, Guinea Pig, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.