Learn More
DDX5 Monoclonal Antibody (Ab204), Alexa Fluor™ 647, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$504.00
Specifications
| Antigen | DDX5 |
|---|---|
| Clone | Ab204 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
51-600-542
![]()
|
Invitrogen™
51600542 |
100 Tests |
Each of 1 for $504.00
|
|
|||||
Description
Description: This Ab204 monoclonal antibody detects human DDX5 protein also known as DEAD box protein 5 and RNA helicase p68. This antibody has been reported to cross-react with mouse and rat orthologs of DDX5 protein. Applications Reported: This Ab204 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This Ab204 antibody has been pre-diluted and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins located at www.thermofisher.com/flowprotocols. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Applications Reported: This Ab204 antibody has been reported for use in immunocytochemical staining. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a RNA-dependent ATPase, and also a proliferation-associated nuclear antigen, specifically reacting with the simian virus 40 tumor antigen. This gene consists of 13 exons, and alternatively spliced transcripts containing several intron sequences have been detected, but no isoforms encoded by these transcripts have been identified.Specifications
| DDX5 | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Human | |
| 1655 | |
| Purified T antigen-related protein from Ad2+ ND2 virus-infected HeLa cells. | |
| Primary | |
| Antibody |
| Ab204 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P17844 | |
| DDX5 | |
| IgG1 κ | |
| 4°C, store in dark, DO NOT FREEZE! | |
| DDX5 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.