Learn More
Description
Specifications
Specifications
| Antigen | DDX56 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | ATP-dependent 61 kDa nucleolar RNA helicase, DDX26, DEAD (Asp-Glu-Ala-Asp) box polypeptide 56, DEAD box protein 21, DEAD box protein 56, DEAD-box RNA helicase, EC 3.6.1, EC 3.6.4.13,61-kd nucleolar helicase, NOH61DDX21, probable ATP-dependent RNA helicase DDX56, putative nucleolar RNA helicase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human DDX56 fragment. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
