Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX56 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB102496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102496 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102496 Supplier Novus Biologicals Supplier No. NBP297523AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX56 Polyclonal specifically detects DDX56 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DDX56
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias ATP-dependent 61 kDa nucleolar RNA helicase, DDX26, DEAD (Asp-Glu-Ala-Asp) box polypeptide 56, DEAD box protein 21, DEAD box protein 56, DEAD-box RNA helicase, EC 3.6.1, EC 3.6.4.13,61-kd nucleolar helicase, NOH61DDX21, probable ATP-dependent RNA helicase DDX56, putative nucleolar RNA helicase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DDX56 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54606
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.