Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX56 Antibody (OTI2E5), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N271964A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271964A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271964A647 Supplier Novus Biologicals Supplier No. NBP271964AF647

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

DDX56 Monoclonal specifically detects DDX56 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen DDX56
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2E5
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATP-dependent 61 kDa nucleolar RNA helicase, DDX26, DEAD (Asp-Glu-Ala-Asp) box polypeptide 56, DEAD box protein 21, DEAD box protein 56, DEAD-box RNA helicase, EC 3.6.1, EC 3.6.4.13,61-kd nucleolar helicase, NOH61DDX21, probable ATP-dependent RNA helicase DDX56, putative nucleolar RNA helicase
Gene Symbols DDX56
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 323-547 of human DDX56 (NP_061955) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54606.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.