Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX59 Antibody (OTI2G1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288294 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288294 Supplier Novus Biologicals Supplier No. NBP272229APC

Mouse Monoclonal Antibody

DDX59 Monoclonal antibody specifically detects DDX59 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen DDX59
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G1
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 59, DEAD box protein 59, DKFZP564B1023, EC 3.6.1, EC 3.6.4.13, Zinc finger HIT domain-containing protein 5, ZNHIT5probable ATP-dependent RNA helicase DDX59
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-303 of human DDX59 (NP_001026895) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83479
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.