Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX60 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342699 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342699 Supplier Novus Biologicals Supplier No. NBP281712MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX60 Polyclonal antibody specifically detects DDX60 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen DDX60
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 60, DEAD box protein 60
Host Species Rabbit
Immunogen DDX60 antibody was raised against a 17 amino acid synthetic peptide near the center of human DDX60. The immunogen is located within amino acids 1140 - 1190 of DDX60. Amino Acid Squence: LKKKQETKRPPKADKEA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 55601
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.