Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEAF1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP214929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214929 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214929 Supplier Novus Biologicals Supplier No. NBP214929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEAF1 Polyclonal specifically detects DEAF1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DEAF1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunoprecipitation 1:10 - 1:500
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O75398
Gene Alias deformed epidermal autoregulatory factor 1 (Drosophila), Nuclear DEAF-1-related transcriptional regulator, NUDRdeformed epidermal autoregulatory factor 1 homolog, SPNsuppressin, Zinc finger MYND domain-containing protein 5, ZMYND5Suppressin
Gene Symbols DEAF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DEAF1. The exact sequence is proprietary.
Molecular Weight of Antigen 59 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10522
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.