Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Death Ligand Signal Enhancer Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341899 Supplier Novus Biologicals Supplier No. NBP281912MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Death Ligand Signal Enhancer Polyclonal antibody specifically detects Death Ligand Signal Enhancer in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Death Ligand Signal Enhancer
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DAP3 Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, Long Form, Death Ligand Signal Enhancer, DELE, DELE1, DELE1(L), hypothetical protein LOC9812, KIAA0141
Host Species Rabbit
Immunogen Death Ligand Signal Enhancer antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human Death Ligand Signal Enhancer. The immunogen is located within the first 50 amino acids of Death Ligand Signal Enhancer. Amino Acid Squence: RVTPKSTSPDGPQTTSST
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9812
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.