Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC-205/CD205 Antibody (019), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB388713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388713 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388713 Supplier Novus Biologicals Supplier No. NBP312780AF350

Rabbit Monoclonal Antibody

DEC-205/CD205 Monoclonal antibody specifically detects DEC-205/CD205 in Mouse samples. It is validated for Western Blot

Specifications

Antigen DEC-205/CD205
Applications Western Blot
Classification Monoclonal
Clone 019
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CD205;DCL-1 fusion;DEC-205;gp200;Ly-75;Lymphocyte antigen 75;membrane antigen MN;RP23-10L17.4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Mouse DEC-205/CD205
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.