Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC-205/CD205 Antibody (MG38), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP2003289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2003289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2003289 Supplier Novus Biologicals Supplier No. NBP200328IR0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen DEC-205/CD205
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MG38
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen The immunogen for this antibody is CD205 (DEC-205).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.