Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC-205/CD205 Antibody (NLDC-145), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. DDX0020A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0020A488 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0020A488 Supplier Novus Biologicals Supplier No. DDX0020A488100
Add to Cart
Add to Cart

Rat Monoclonal Antibody

DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DEC-205/CD205
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone NLDC-145
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Accession No. NP_005840
Gene Symbols LY75
Host Species Rat
Immunogen Dec205/CD205 (NLDC145) was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.