Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC-205/CD205 Mouse anti-Human, Primate, DyLight™ 650, Clone: HD30, Novus Biologicals™
SDP

Catalog No. NBP141003C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141003C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141003C Supplier Novus Biologicals Supplier No. NBP141003C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen DEC-205/CD205
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HD30
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Symbols LY75
Host Species Mouse
Immunogen Purified extracellular domain of human CD205-IgG1 Fc fusion protein.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary or Secondary Primary
Test Specificity This antibody specifically recognises CD205, otherwise known as DEC205, a type I transmembrane glycoprotein and member of the macrophage mannose receptor family of C-type lectins, predominantly expressed by human dendritic cells.
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.