Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ DEC-205/CD205 Recombinant Protein Antigen
Shop All R&D Systems Products

Click to view available options
Quantity:
100 μL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Renal Cell Carcinoma (gp200). Source: E.coli Amino Acid Sequence: QRHRLHLAGFSSVRYAQGVNEDEIMLPSFH The DEC-205/CD205 Recombinant Protein Antigen is derived from E. coli. The DEC-205/CD205 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Gene ID (Entrez) | 4065 |
Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
Common Name | Renal Cell Carcinoma (gp200) Recombinant Protein Antigen |
Content And Storage | Store at −20C. Avoid freeze-thaw cycles. |
Formulation | PBS and 1M Urea, pH 7.4. |
For Use With (Application) | Blocking/Neutralizing, Control |
Gene Alias | CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75 |
Gene Symbol | LY75 |
Label Type | Unlabeled |
Product Type | Recombinant Protein Antigen |
Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction