Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB1001800H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001800H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001800H Supplier Novus Biologicals Supplier No. NB1001800H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEC1 Polyclonal specifically detects DEC1 in Human, Mouse, Rat samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen DEC1
Applications ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting
Formulation PBS with No Preservative
Gene Accession No. O14503
Gene Alias basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14
Gene Symbols BHLHE40
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 8553
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.