Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB213255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB213255 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB213255 Supplier Novus Biologicals Supplier No. NB1001800PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEC1. Polyclonal specifically detects DEC1. in Human, Mouse, Rat samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen DEC1.
Applications ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 8553
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.