Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC2/SHARP1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP119613A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP119613A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP119613A94 Supplier Novus Biologicals Supplier No. NBP119613AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEC2/SHARP1 Polyclonal specifically detects DEC2/SHARP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DEC2/SHARP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2
Gene Symbols BHLHE41
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human DEC2/SHARP1 inbetween residues 1-75 [UniProt Q9C0J9].
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79365.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.