Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC2/SHARP1 Antibody (OTI5B1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272203G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272203G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272203G Supplier Novus Biologicals Supplier No. NBP272203G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DEC2/SHARP1 Monoclonal specifically detects DEC2/SHARP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen DEC2/SHARP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5B1
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2
Gene Symbols BHLHE41
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-297 of human DEC2/SHARP1 (NP_110389) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79365.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.