Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decorin Antibody (DCN/6289), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB278922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB278922 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB278922 Supplier Novus Biologicals Supplier No. NBP321077MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Decorin Monoclonal antibody specifically detects Decorin in Human, Mouse, Porcine, Bovine, Canine, Chicken, Equine, Elk, Rabbit samples. It is validated for Flow Cytometry

Specifications

Antigen Decorin
Applications Flow Cytometry
Classification Monoclonal
Clone DCN/6289
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B
Host Species Mouse
Immunogen Recombinant full-length Decorin protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1634
Target Species Human, Mouse, Pig, Bovine, Canine, Chicken, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.