Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DECR2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP153172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP153172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP153172 Supplier Novus Biologicals Supplier No. NBP153172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DECR2 Polyclonal specifically detects DECR2 in Human samples. It is validated for Western Blot.

Specifications

Antigen DECR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NUI1
Gene Alias 2,4-dienoyl CoA reductase 2, peroxisomal, EC 1.3.1, EC 1.3.1.34, pDCR2,4-dienoyl-CoA reductase 2, PDCRshort chain dehydrogenase/reductase family 17C, member 1, peroxisomal 2,4-dienoyl-CoA reductase, SDR17C1
Gene Symbols DECR2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DECR2(2,4-dienoyl CoA reductase 2, peroxisomal) The peptide sequence was selected from the N terminal of DECR2. Peptide sequence MAQPPPDVEGDDCLPAYRHLFCPDLLRDKVAFITGGGSGIGFRIAEIFMR.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 26063
Test Specificity Expected identity based on immunogen sequence: Xenopus: 100%; Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.