Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DECR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | DECR2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DECR2 Polyclonal specifically detects DECR2 in Human samples. It is validated for Western Blot.Specifications
DECR2 | |
Polyclonal | |
Purified | |
RUO | |
Q9NUI1 | |
26063 | |
Synthetic peptides corresponding to DECR2(2,4-dienoyl CoA reductase 2, peroxisomal) The peptide sequence was selected from the N terminal of DECR2. Peptide sequence MAQPPPDVEGDDCLPAYRHLFCPDLLRDKVAFITGGGSGIGFRIAEIFMR. | |
Primary |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
2,4-dienoyl CoA reductase 2, peroxisomal, EC 1.3.1, EC 1.3.1.34, pDCR2,4-dienoyl-CoA reductase 2, PDCRshort chain dehydrogenase/reductase family 17C, member 1, peroxisomal 2,4-dienoyl-CoA reductase, SDR17C1 | |
DECR2 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title