Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-1/CLEC7A Antibody (003), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB333045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333045 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333045 Supplier Novus Biologicals Supplier No. NBP290490JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Dectin-1/CLEC7A Monoclonal antibody specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for ELISA

Specifications

Antigen Dectin-1/CLEC7A
Applications ELISA
Classification Monoclonal
Clone 003
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Dectin-1/CLEC7A (Accession#: NP_064392.2, Phe69-Leu244)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64581
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.