Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-1/CLEC7A Antibody (15E2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB118930 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB118930 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB118930 Supplier Novus Biologicals Supplier No. NBP307900IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Dectin-1/CLEC7A
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1500
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Host Species Mouse
Immunogen Extracellular part of human CD369 with hIgG FC tag
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64581
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.