Learn More
Description
Specifications
Specifications
| Antigen | Dectin-1/CLEC7A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BD6 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry |
| Gene Accession No. | Q9BXN2 |
| Gene Alias | Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 |
| Gene Symbols | CLEC7A |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
