Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-2/CLEC6A Antibody (3D1), PE, Novus Biologicals™
SDP

Catalog No. NBP227159PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227159PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227159PE Supplier Novus Biologicals Supplier No. NBP227159PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Dectin-2/CLEC6A
Applications Flow Cytometry
Classification Monoclonal
Clone 3D1
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. Q6EIG7
Gene Alias CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Gene Symbols CLEC6A
Host Species Mouse
Immunogen A portion of amino acids 75-139 of human Dectin-2 was used as the immunogen for the antibody. 90-106
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93978
Test Specificity The amino acids sequence used as immunogen is 100% homologous in Human, Orangutan and Chimp, 83% homologous in Rhesus Monkey, Bovine and Sheep, 65% homologous in Mouse and 52% homologous in Rat.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.