Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-2/CLEC6A Antibody (545925), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB363301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363301 Supplier Novus Biologicals Supplier No. FAB31141W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Dectin-2/CLEC6A
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 545925
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Dectin-2/CLEC6A, Thr46-Leu209, Accession # Q6EIG7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93978
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.