Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-2/CLEC6A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen Dectin-2/CLEC6A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15973920
SDP
View Documents
Novus Biologicals
NBP15973920UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159739
SDP
View Documents
Novus Biologicals
NBP159739
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Dectin-2/CLEC6A Polyclonal specifically detects Dectin-2/CLEC6A in Human samples. It is validated for Western Blot.
Specifications

Specifications

Dectin-2/CLEC6A
Polyclonal
Rabbit
Q6EIG7
93978
Synthetic peptides corresponding to CLEC6A(C-type lectin domain family 6, member A) Antibody(against the N terminal of CLEC6A (NP_001007034). Peptide sequence FIVSCVVTYHFTYGETGKRLSELHSYHSSLTCFSEGTKVPAWGCCPASWK.
Primary
Western Blot
Unconjugated
RUO
CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
CLEC6A
IgG
24 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.