Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-2/CLEC6A Antibody (RM0220-7B45), PerCP, Novus Biologicals™
SDP

Catalog No. NB361156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361156 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361156 Supplier Novus Biologicals Supplier No. NBP212260PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Dectin-2/CLEC6A Monoclonal antibody specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for Western Blot

Specifications

Antigen Dectin-2/CLEC6A
Applications Western Blot
Classification Monoclonal
Clone RM0220-7B45
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Host Species Rat
Immunogen Mouse recombinant Dectin-2/CLEC6A
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93978
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.