Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals

Catalog No. p-7043236
Click to view available options
Quantity:
1 g

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58846-77-8
Molecular Formula C16H32O6
Molecular Weight (g/mol) 320.426
MDL Number MFCD00063297
InChI Key JDRSMPFHFNXQRB-IBEHDNSVSA-N
Synonym decyl beta-d-glucopyranoside, n-decyl-b-d-glucopyranoside, decyl-beta-d-glucopyranoside, n-decyl-beta-d-glucopyranoside, unii-z17h97ea6y, n-decyl glucoside, decyl beta-d-glucoside, triton x 190, 28-deacetylsendanin, beta-d-glucoside, decyl
PubChem CID 62142
IUPAC Name (2R,3R,4S,5S,6R)-2-decoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 1 g
Beilstein 85089
Solubility Information Soluble in methanol
Formula Weight 320.42
Physical Form Powder
Chemical Name or Material Decyl beta-D-glucopyranoside
EINECSNumber 261-469-7
RTECSNumber LZ5776531
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.