Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEDD Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726622 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-172-6622 150 μL
1 options

Catalog No. 50-172-6622

Supplier: Proteintech Group Inc 108161AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEDD encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen DEDD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DEDD
Gene Accession No. O75618
Gene Alias CASP8IP1, DEDD, DEDD1, DEDPro1, DEFT, FLDED 1, FLDED1, KE05
Gene Symbols DEDD
Host Species Rabbit
Immunogen DEDD Fusion Protein Ag1195
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9191
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.