Learn More
Description
Specifications
Specifications
| Antigen | DEDD2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | death effector domain containing 2, death effector domain-containing DNA binding protein 2, DED-containing protein FLAME-3, DNA-binding death effector domain-containing protein 2, FADD-like apoptotic molecule 3, FLAME3, FLAME-3 |
| Gene Symbols | DEDD2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 175-193 (APAAPQQQSEPARPSSEGK) of human DEDD2/Flame-3 was used as immunogen; GenBank no. gi 19923050 ref NP_579874.1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
