Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dedicator of cytokinesis 8 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017395 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017395 Supplier Novus Biologicals Supplier No. NBP282084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

dedicator of cytokinesis 8 Polyclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen dedicator of cytokinesis 8
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias dedicator of cytokinesis 8, FLJ00026, FLJ00346
Gene Symbols DOCK8
Host Species Rabbit
Immunogen Dedicator of cytokinesis 8 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human dedicator of cytokinesis 8. The immunogen is located within the last 50 amino acids of dedicator of cytokinesis 8. Amino Acid Squence: RVESQKRDSFHRSSFRK
Molecular Weight of Antigen 220, 230 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81704
Test Specificity dedicator of cytokinesis 8 antibody is human, mouse and rat reactive. Multiple isoforms of dedicator of cytokinesis 8 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.