Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dedicator of cytokinesis 8 Antibody (OTI12D7), FITC, Novus Biologicals™
SDP

Catalog No. NBP272227F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272227F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272227F Supplier Novus Biologicals Supplier No. NBP272227F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen dedicator of cytokinesis 8
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI12D7
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias dedicator of cytokinesis 8, FLJ00026, FLJ00346
Gene Symbols DOCK8
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 84-405 of human dedicator of cytokinesis 8 (NP_982272) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81704.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.