Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dedicator of cytokinesis 8 Antibody (OTI4F3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272226J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272226J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272226J549 Supplier Novus Biologicals Supplier No. NBP272226JF549

Mouse Monoclonal Antibody

dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen dedicator of cytokinesis 8
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI4F3
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dedicator of cytokinesis 8, FLJ00026, FLJ00346
Gene Symbols DOCK8
Host Species Mouse
Immunogen Full length recombinant protein of human dedicator of cytokinesis 8 (NP_982272) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81704.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.