Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dedicator of cytokinesis 8 Antibody (OTI4H10), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272228A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272228A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272228A94 Supplier Novus Biologicals Supplier No. NBP272228AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen dedicator of cytokinesis 8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4H10
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias dedicator of cytokinesis 8, FLJ00026, FLJ00346
Gene Symbols DOCK8
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 832-1160 of human dedicator of cytokinesis 8 (NP_982272) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81704
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.