Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ DNA LoBind™ Plates
GREENER_CHOICE

Catalog No. 05414216 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Blue
White
White, Clear
White, Clear Bottom
No. of Wells:
96
384
Volume (Metric) Well:
140 μL
200 μL
350 μL
500 μL
1000 μL
Packaging:
10 Bags x 12 Plates
10 Bags x 8 Plates
5 Bags x 16 Plates
5 Bags x 4 Plates
5 Bags x 8 Plates
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells Volume (Metric) Well Packaging
05-414-216 White, Clear 96 350 μL 5 Bags x 16 Plates
E951032000 White 96 500 μL 5 Bags x 8 Plates
05-414-229 Blue 96 500 μL 5 Bags x 8 Plates
E951032409 White, Clear Bottom 96 500 μL 10 Bags x 12 Plates
05-414-238 Blue 96 1000 μL 5 Bags x 4 Plates
E951032808 White 96 1000 μL 5 Bags x 4 Plates
E951033201 White, Clear Bottom 96 1000 μL 10 Bags x 8 Plates
E951040546 White 384 140 μL 5 Bags x 16 Plates
E951031208 White 384 200 μL 5 Bags x 8 Plates
05-414-221 White 384 200 μL 10 Bags x 12 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 05-414-216 Supplier Eppendorf™ Supplier No. 0030603303
Add to Cart
Add to Cart
This item is not returnable. View return policy

DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing.
TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 16 Plates
Color White, Clear
Product Type Consumables
Material Polypropylene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 20 to 300 μL
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 80 Pk.
Well Color Colorless
Dimensions (L x W x H) 85.5 x 127.8 x 14.4 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.