Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Deepwell Plates 96-well, 1000 μL

Catalog No. 05414236 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Blue
Green
White
White, Clear Bottom
Yellow
Purity or Quality Grade:
PCR Clean (Non-sterile)
Sterile
Packaging:
10 Bags x 8 Plates
5 Bags x 4 Plates
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade Packaging
05-414-236 Green Sterile 5 Bags x 4 Plates
E951032603 White PCR Clean (Non-sterile) 5 Bags x 4 Plates
E951032662 Green PCR Clean (Non-sterile) 5 Bags x 4 Plates
E951032689 Blue PCR Clean (Non-sterile) 5 Bags x 4 Plates
05-414-234 Yellow PCR Clean (Non-sterile) 5 Bags x 4 Plates
E951033006 White PCR Clean (Non-sterile) 10 Bags x 8 Plates
E951032701 White Sterile 5 Bags x 4 Plates
05-414-235 Yellow Sterile 5 Bags x 4 Plates
05-414-237 Blue Sterile 5 Bags x 4 Plates
E951033103 White, Clear Bottom Sterile 10 Bags x 8 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 05-414-236 Supplier Eppendorf™ Supplier No. 951032760
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eppendorf Deepwell Plates offer efficient sample handling with features like OptiTrack™ matrix for rapid identification and g-Safe™ technology for stability during centrifugation.

  • OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling.
  • RecoverMax™ well design: Optimized well geometry for maximum sample recovery and excellent mixing properties.
  • g-Safe™: exceptional centrifugation stability up to 6,000 x g for faster protocols and improved sample quality.
  • Raised well rims and smooth surface for reliable closing, even with heat sealing.
  • Easy and reliable stacking.
  • Minimized remaining volume and high uniformity from well to well.
  • High-quality polypropylene ensures high resistance to chemicals, mechanical stress and temperature extremes.
  • Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays.
  • Autoclavable (121°C, 20 min.).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes

Compliance

IVD, SBS/ANSI™

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color Green
Product Type Deepwell Plate
Material Polypropylene
Volume (Metric) Well 1000 μL
No. of Wells 96
Sterility Sterile
Max. RCF 5000 x g
Purity or Quality Grade Sterile
Quantity 20 Pk.
Well Color Colorless
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.