Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEFB118 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB331232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331232 Supplier Novus Biologicals Supplier No. NBP297548MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEFB118 Polyclonal antibody specifically detects DEFB118 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen DEFB118
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias beta 18, beta-defensin 118, C20orf63, DEFB18, DEFB-18, defensin, beta 118, ESC42, ESP13.6
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DEFB118 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 117285
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.