Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Defensin alpha 5 Antibody (8C8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013766 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013766 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013766 Supplier Novus Biologicals Supplier No. NBP280693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.

Specifications

Antigen Defensin alpha 5
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8C8
Concentration 2.35 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Flow Cytometry : 1 ug per million cells, ELISA reported in scientific literature, Immunohistochemistry : 5-10 ug/ml, Immunocytochemistry/Immunofluorescence : 1:200-1:1000, Immunohistochemistry-Paraffin : 5-10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 30612153), Dot Blot reported in scientific literature
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728
Gene Symbols DEFA5
Host Species Mouse
Immunogen Full-length human Defensin alpha 5 [Swiss-Prot# Q01523]. The propeptide is the full-length protein. In the mature form, it is cleaved at residue 63 (alanine).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1670
Test Specificity Human.
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.