Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Defensin alpha 5 Antibody (8C8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB11060002U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060002U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060002U Supplier Novus Biologicals Supplier No. NB11060002UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Defensin alpha 5
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 8C8
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. Q01523
Gene Alias DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728
Gene Symbols DEFA5
Host Species Mouse
Immunogen Full-length human Defensin alpha 5 [Swiss-Prot# Q01523]. The propeptide is the full-length protein. In the mature form, it is cleaved at residue 63 (alanine).
Molecular Weight of Antigen 10 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1670
Test Specificity This is specific for HD5. It does not cross-react with HD6, lysozyme or sPLA2.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.