Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEGS2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151790 25 μg
NB151789 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151790 Supplier Novus Biologicals Supplier No. NBP31191825UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEGS2 Polyclonal antibody specifically detects DEGS2 in Human samples. It is validated for Western Blot

Specifications

Antigen DEGS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400
Formulation PBS
Gene Alias C14orf66, chromosome 14 open reading frame 66, Degenerative spermatocyte homolog 2, degenerative spermatocyte homolog 2, lipid desaturase (Drosophila), DES2sphingolipid delta(4)-desaturase/C4-hydroxylase DES2, EC 1.14, FADS8, sphingolipid C4-hydroxylase/delta 4-desaturase, sphingolipid delta 4 desaturase/C-4 hydroxylase
Host Species Rabbit
Immunogen Synthetic peptide derived from human DEGS2 protein. (Uniprot: Q6QHC5)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 123099
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.