Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Dehydrated Culture Media and Ingredients
Shop All Fischer Technical Company Products

Click to view available options
Media Type:
Agar Powder
MacConkey Agar
Nutrient Agar
Nutrient Agar 1.5%
Nutrient Broth
Potato Dextrose Agar
Sabouraud Dextrose Agar
Starch Agar
Tryptic Soy Agar
Quantity:
100 g
113 g
454 g
500 g
Description
Successful bacteriological work begins with high-quality media.
- Agar, a solidifying agent, is an essential component of many media
- Available in a variety of formulations and bottle sizes
- For use in preparing plated media, tubed media, bottled media, and slants
Specifications
Specifications
Form | Dehydrated Powder |
Format | Bottled |
Manufacturing Origin | US with Globally Sourced Materials |
Media Type | Starch Agar |
Product Type | Microbiology Media |
Sterility | Non-sterile |
Target Organism Class | Bacteria |
Test Type | Microbiological Tests |
For Use With (Application) | Microbiological Testing |
Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction