Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Dehydrated Culture Media and Ingredients
Shop All Fischer Technical Company Products

Click to view available options
Media Type:
Agar Powder
MacConkey Agar
Nutrient Agar
Nutrient Agar 1.5%
Nutrient Broth
Potato Dextrose Agar
Sabouraud Dextrose Agar
Starch Agar
Tryptic Soy Agar
Quantity:
100 g
113 g
454 g
500 g
Description
Successful bacteriological work begins with high-quality media.
- Agar, a solidifying agent, is an essential component of many media
- Available in a variety of formulations and bottle sizes
- For use in preparing plated media, tubed media, bottled media, and slants
Specifications
Specifications
Form | Dehydrated |
Format | Bottled |
Manufacturing Origin | US with Globally Sourced Materials |
Media Type | Nutrient Agar 1.5% |
Product Type | Microbiology Media |
Sterility | Non-sterile |
Target Organism Class | Bacteria |
Test Type | Microbiological Tests |
For Use With (Application) | Microbiological Testing |
Quantity | 454 g |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction