Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrin Antibody, Novus Biologicals™
SDP

Catalog No. NB036581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036581 Supplier Novus Biologicals Supplier No. NBP288990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dehydrin Polyclonal specifically detects Dehydrin in Plant samples. It is validated for Western Blot.

Specifications

Antigen Dehydrin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 50% glycerol
Gene Alias RAB21, Water stress-inducible protein Rab21
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence near the C-terminus of Dehydrin. The sequence is completely conserved in barley, rice, maize and wheat.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.